4-Fluoro-7-nitrobenzofurazan

Supplier: Matrix Scientific

Synonyms: NBD-F4-Fluoro-7-nitro-[2,1,3]-benzoxadiazole

002807-100MG
101784-702EA 500 USD
101784-702
4-Fluoro-7-nitrobenzofurazan
4-Fluoro-7-nitrobenzofurazan
Formula: C₆H₂FN₃O₃
MW: 183.1 g/mol
MDL Number: MFCD00010196
CAS Number: 29270-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR