2-Chloro-6-fluoro-m-toluic acid

Supplier: Matrix Scientific

Synonyms: 2-Chloro-6-fluoro-3-methylbenzoic acid

002755-1G 002755-5G
101784-596EA 44.62 USD
101784-596 101784-598
2-Chloro-6-fluoro-m-toluic acid
2-Chloro-6-fluoro-m-toluic acid
Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD01631359
CAS Number: 32890-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR