2-Fluoro-4-nitrotoluene ≥98%

Supplier: Matrix Scientific

002598-25G 002598-100G
101784-256EA 20 USD
101784-256 101784-258
2-Fluoro-4-nitrotoluene ≥98%
2-Fluoro-4-nitrotoluene

Hazard code(s): Irritant

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
Boiling Pt: 65…68 °C (2mm)
Melting Pt: 34…37 °C
Storage Temperature: Ambient
MDL Number: MFCD00007199
CAS Number: 1427-07-2
EINECS: 215-845-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 65-68°C/2mm
Melting point 34-37°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR