3-Fluoro-4-nitrophenol ≥99%
Supplier: Matrix Scientific
|
Warning
|
002595-25G
002595-5G
101784-244EA
33.85
USD
101784-244
101784-246
3-Fluoro-4-nitrophenol ≥99%
3-Fluoro-4-nitrophenol
Hazard code(s): Irritant
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Melting Pt: 93…95 °C Storage Temperature: Ambient |
MDL Number:
MFCD00041251 CAS Number: 394-41-2 EINECS: 206-895-6 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...