3-Fluoro-4-methylbenzyl alcohol

Supplier: Matrix Scientific

Synonyms: (3-Fluoro-4-methylphenyl)methanol

002576-1G 002576-5G
101784-194EA 76.92 USD
101784-194 101784-196
3-Fluoro-4-methylbenzyl alcohol
3-Fluoro-4-methylbenzyl alcohol
Formula: C₈H₉FO
MW: 140.16 g/mol
MDL Number: MFCD01631533
CAS Number: 192702-79-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR