2-Chloro-4-iodotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-iodo-1-methylbenzene1-Chloro-5-iodo-2-methylbenzene

002552-25G
101784-148EA 110.77 USD
101784-148
2-Chloro-4-iodotoluene ≥98%
2-Chloro-4-iodotoluene

Hazard code(s): Irritant

Formula: C₇H₆ClI
MW: 252.48 g/mol
Boiling Pt: 145 °C (35mm)
Density: 1.80 g/cm³
MDL Number: MFCD00040872
CAS Number: 83846-48-4
EINECS: 281-024-0

Order Now

Specification Test Results

Boiling point 145°C/35mm
Density 1.8
ND20 1.6211

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR