Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Fluoro-4-iodobenzene ≥99%

Supplier: Matrix Scientific

Warning
Synonyms: 4-Fluoroiodobenzene

002547-100G
101784-132EA 61.46 USD InStock
101784-132
1-Fluoro-4-iodobenzene ≥99%
1-Fluoro-4-iodobenzene
(Stabilized with copper)

n20/D: 1.5830
Hazard code(s): Irritant


Formula: C₆H₄FI
MW: 222 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: -18 °C
Density: 1.9253 g/cm³ (15 °C)
MDL Number: MFCD00001052
CAS Number: 352-34-1
EINECS: 206-522-7
Order Now
Specification Test Results
Boiling point 182-184°C
Melting point -20°C
Density 1.925
ND20 1.583


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty