2-Chloro-4-fluorophenol ≥99%

Supplier: Matrix Scientific
Warning

002521-100G
101784-048EA 92.31 USD
101784-048
2-Chloro-4-fluorophenol ≥99%
2-Chloro-4-fluorophenol

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 88 °C (4mm)
Melting Pt: 23 °C
Density: 1.344 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00002168
CAS Number: 1996-41-4
EINECS: 217-876-7
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Boiling point 88°C/4mm
Melting point 23°C
Density 1.344
ND20 1.53

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR