3-Chloro-4-fluorophenol ≥98%

Supplier: Matrix Scientific

002516-25G 002516-100G
101784-030EA 58.46 USD
101784-030 101784-032
3-Chloro-4-fluorophenol ≥98%
3-Chloro-4-fluorophenol

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₆H₄ClFO
MW: 146.55 g/mol
Boiling Pt: 225 °C (1013 hPa)
Melting Pt: 42…44 °C
MDL Number: MFCD00002257
CAS Number: 2613-23-2
EINECS: 220-042-5
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Melting point 42-44°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR