2-Chloro-4-(trifluoromethyl)benzonitrile ≥98%
Supplier: Matrix Scientific
002481-5G
002481-25G
101783-960EA
21.54
USD
101783-960
101783-962
2-Chloro-4-(trifluoromethyl)benzonitrile ≥98%
2-Chloro-4-(trifluoromethyl)benzonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₈H₃ClF₃N MW: 205.57 g/mol Boiling Pt: 82…83 °C (8mm) |
MDL Number:
MFCD00084939 CAS Number: 1813-33-8 UN: 3276 ADR: 6.1,III |
Specification Test Results
Boiling point | 82-83°C/8mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...