4-Fluoro-3-methylbenzaldehyde
Supplier: Matrix Scientific
Synonyms:
4-Fluoro-m-tolualdehyde
, 4-Fluoro-m-tolylaldehyde
, 4-Fluoro-3-tolualdehyde
002373-25G
002373-5G
101783-790EA
100
USD
101783-790
101783-788
4-Fluoro-3-methylbenzaldehyde
4-Fluoro-3-methylbenzaldehyde
Formula:
C₈H₇FO MW: 138.14 g/mol |
MDL Number:
MFCD01631431 CAS Number: 135427-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...