1-Chloro-3-iodobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Chloroiodobenzene

002356-100G
101783-736EA 75.38 USD
101783-736
1-Chloro-3-iodobenzene ≥98%
1-Chloro-3-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₄ClI
MW: 238.45 g/mol
Boiling Pt: 103…104 °C (15mm)
Melting Pt: 54.5 °C
Density: 1.93 g/cm³
MDL Number: MFCD00001046
CAS Number: 625-99-0
EINECS: 210-920-6

Order Now

Specification Test Results

Boiling point 103-104°C/1
Density 1.93
ND20 1.631

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR