4-Chloro-3-(trifluoromethyl)benzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-α,α,α-trifluoro-m-toluic acid

002305-5G
101783-650EA 40 USD
101783-650
4-Chloro-3-(trifluoromethyl)benzoic acid ≥98%
4-Chloro-3-(trifluoromethyl)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD01631468
CAS Number: 1737-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR