(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-Fluoro-3-(trifluoromethyl)phenylacetic acid
002291-1G
002291-5G
101783-620EA
218.46
USD
101783-620
101783-618
(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid ≥98%
(2-Fluoro-3-(trifluoromethyl)phenyl)acetic acid
Hazard code(s): Irritant
Formula:
C₉H₆F₄O₂ MW: 222.14 g/mol |
MDL Number:
MFCD00061185 CAS Number: 194943-83-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...