2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
1-Chloro-2,4-dinitro-6-(trifluoromethyl)benzene
, 2-Chloro-1,5-dinitro-3-(trifluoromethyl)benzene
, 2-Chloro-3,5-dinitrobenzotrifluoride
002259-25G
002259-100G
101783-554EA
250.77
USD
101783-554
101783-552
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene ≥97%
2-Chloro-3,5-dinitro-α,α,α-trifluorotoluene
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₂ClF₃N₂O₄ MW: 270.55 g/mol Melting Pt: 62…64 °C |
MDL Number:
MFCD00007076 CAS Number: 392-95-0 EINECS: 206-883-0 UN: 2923 ADR: 8,III |
Specification Test Results
Melting point | 62-64°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...