1H,1H,2H,2H-Heneicosafluoro-1-dodecanol ≥97%
Supplier: Matrix Scientific
Synonyms:
3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Heneicosafluoro-1-dodecanol
, 1H,1H,2H,2H-Perfluoro-1-dodecanol
, 1H,1H,2H,2H-Perfluorododecan-1-ol
002223-5G
101783-490EA
403.08
USD
101783-490
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol ≥97%
1H,1H,2H,2H-Heneicosafluoro-1-dodecanol
Hazard code(s): Irritant
Formula:
C₁₂H₅F₂₁O MW: 564.14 g/mol Boiling Pt: 110…112 °C (10mm) Melting Pt: 92…93 °C |
MDL Number:
MFCD00039545 CAS Number: 865-86-1 EINECS: 212-748-7 |
Specification Test Results
Boiling point | 110-112°C/1 |
Melting point | 92-93°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...