5-Fluoro-2-nitrophenol ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Fluoro-2-hydroxynitrobenzene
002196-250G
002196-100G
002196-25G
101783-452EA
273.85
USD
101783-452
101783-450
101783-448
5-Fluoro-2-nitrophenol ≥97%
5-Fluoro-2-nitrophenol
Hazard code(s): Irritant
Formula:
C₆H₄FNO₃ MW: 157.1 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 35…37 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007107 CAS Number: 446-36-6 EINECS: 207-168-6 |
Specification Test Results
Melting point | 35-37°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...