Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Fluoro-2-nitroaniline ≥97%

Supplier: Matrix Scientific


002189-100G 002189-25G
101783-430EA 57.54 USD InStock
101783-430 101783-428
4-Fluoro-2-nitroaniline ≥97%
4-Fluoro-2-nitroaniline
Hazard code(s): Irritant


Formula: C₆H₅FN₂O₂
MW: 156.12 g/mol
Melting Pt: 95 °C
Storage Temperature: Ambient
MDL Number: MFCD00007830
CAS Number: 364-78-3
EINECS: 206-666-0
Order Now
Specification Test Results
Melting point 91-93°C


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty