4-Chloro-2-fluoroaniline ≥98%

Supplier: Matrix Scientific

002082-25G 002082-100G
101783-166EA 69.23 USD
101783-166 101783-164
4-Chloro-2-fluoroaniline ≥98%
4-Chloro-2-fluoroaniline

Hazard code(s): Toxic

Formula: C₆H₅ClFN
MW: 145.56 g/mol
Boiling Pt: 92 °C (2mm)
Density: 1.311 g/cm³
MDL Number: MFCD00010625
CAS Number: 57946-56-2
EINECS: 261-034-1
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Density 1.311
ND20 1.56

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR