4-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: Matrix Scientific

Synonyms: (4-Fluoro-2-(trifluoromethyl)phenyl)methanol

001982-5G 001982-25G
101782-990EA 76.92 USD
101782-990 101782-992
4-Fluoro-2-(trifluoromethyl)benzyl alcohol ≥97%
4-Fluoro-2-(trifluoromethyl)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₆F₄O
MW: 194.13 g/mol
Melting Pt: 55…57 °C
MDL Number: MFCD00061170
CAS Number: 220227-29-2

Order Now

Specification Test Results

Melting point 55-57°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR