2-Nitro-4-(trifluoromethyl)benzonitrile ≥98%
Supplier: Matrix Scientific
Synonyms:
4-Cyano-3-nitrobenzotrifluoride
001849-25G
001849-100G
101782-724EA
63.08
USD
101782-724
101782-726
2-Nitro-4-(trifluoromethyl)benzonitrile ≥98%
2-Nitro-4-(trifluoromethyl)benzonitrile
Hazard code(s): Toxic
Formula:
C₈H₃F₃N₂O₂ Boiling Pt: 156…158 °C (18mm) Melting Pt: 44…45 °C |
MDL Number:
MFCD00014684 CAS Number: 778-94-9 EINECS: 212-298-1 UN: 3439 ADR: 6.1,III |
Specification Test Results
Boiling point | 156-158°C/1 |
Melting point | 44-45°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...