4-Nitro-3-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Nitro-α,α,α-trifluoro-m-toluidine5-Amino-2-nitrobenzotrifluoride5-Amino-2-nitro-α,α,α-trifluorotoluene

001846-100G
101782-716EA 63.08 USD
101782-716
4-Nitro-3-(trifluoromethyl)aniline ≥98%
4-Nitro-3-(trifluoromethyl)aniline

Hazard code(s): Irritant-Harmful, Refrigerate

Formula: C₇H₅F₃N₂O₂
MW: 206.12 g/mol
Melting Pt: 126…128 °C
Density: 1.752 g/cm³ (–123.15 °C)
MDL Number: MFCD00014717
CAS Number: 393-11-3
EINECS: 206-884-6
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 126-128°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR