2-Bromo-p-toluic acid ≥98%

Supplier: Matrix Scientific

001816-5G
101782-656EA 73.85 USD
101782-656
2-Bromo-p-toluic acid ≥98%
2-Bromo-p-toluic acid

Hazard code(s): Irritant

Formula: C₈H₇BrO₂
MW: 215.05 g/mol
Melting Pt: 143…145 °C
MDL Number: MFCD00045797
CAS Number: 7697-27-0

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR