1-Bromo-2-iodobenzene ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-Bromo-1-iodobenzene
, 2-Bromoiodobenzene
Stabilization:
Stabilised with copper
001774-25G
001774-100G
101782-542EA
32.31
USD
101782-542
101782-544
1-Bromo-2-iodobenzene ≥97%
1-Bromo-2-iodobenzene
Hazard code(s): Irritant, Light Sensitive
Formula:
C₆H₄BrI MW: 282.91 g/mol Boiling Pt: 120…121 °C (15mm) Melting Pt: 9…10 °C Density: 2.203 g/cm³ |
MDL Number:
MFCD00001030 CAS Number: 583-55-1 EINECS: 209-508-9 |
Specification Test Results
Boiling point | 120-121°C/1 |
Melting point | 9-10°C |
Density | 2.203 |
ND20 | 1.661 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...