5-Bromo-2-fluorotoluene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Bromo-1-fluoro-2-methylbenzene

001758-100G
101782-500EA 30.77 USD
101782-500
5-Bromo-2-fluorotoluene ≥99%
5-Bromo-2-fluorotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 95 °C (50mm)
Density: 1.49 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000343
CAS Number: 51437-00-4
EINECS: 257-202-9

Order Now

Specification Test Results

Boiling point 95°C/50mm
Density 1.49
ND20 1.529

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR