2-Bromo-5-fluorotoluene ≥98%

Supplier: Matrix Scientific
Danger

Synonyms: 1-Bromo-4-fluoro-2-methylbenzene

001755-100G
101782-488EA 173.85 USD
101782-488
2-Bromo-5-fluorotoluene ≥98%
2-Bromo-5-fluorotoluene

Hazard code(s): Irritant

Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 170 °C
Density: 1.49 g/cm³
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 170°C
Density 1.49
ND20 1.526

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR