5-Bromo-2-fluorobenzoic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
001720-25G
001720-5G
101782-398EA
20
USD
101782-398
101782-400
5-Bromo-2-fluorobenzoic acid ≥97%
5-Bromo-2-fluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol |
MDL Number:
MFCD00143423 CAS Number: 146328-85-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...