3-Bromo-2-fluorobenzoic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
001716-25G
001716-5G
101782-380EA
36.92
USD
101782-380
101782-382
3-Bromo-2-fluorobenzoic acid ≥97%
3-Bromo-2-fluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol Melting Pt: 168…170 °C |
MDL Number:
MFCD00665763 CAS Number: 161957-56-8 |
Specification Test Results
Melting point | 168-170°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...