1-Bromo-4-chloro-3-fluorobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
4-Bromo-1-chloro-2-fluorobenzene
001609-25G
001609-100G
101782-084EA
23.08
USD
101782-084
101782-086
1-Bromo-4-chloro-3-fluorobenzene ≥98%
1-Bromo-4-chloro-3-fluorobenzene
Hazard code(s): Irritant
Formula:
C₆H₃BrClF MW: 209.44 g/mol Boiling Pt: 62…64 °C (9mm) Density: 1.710 g/cm³ |
MDL Number:
MFCD00042519 CAS Number: 60811-18-9 |
Specification Test Results
Boiling point | 62-64/9mm |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...