4-Bromo-2-(trifluoromethyl)aniline ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-bromo-α,α,α-trifluorotoluene2-Amino-5-bromobenzotrifluoride4-Bromo-α,α,α-trifluoro-o-toluidine2-Amino-5-bromo-α,α,α-trifluortoluene

001594-100G 001594-25G
101782-060EA 51.75 USD
101782-060 101782-058
4-Bromo-2-(trifluoromethyl)aniline ≥98%
4-Bromo-2-(trifluoromethyl)aniline

Hazard code(s): Irritant, Light Sensitive, Refrigerate

Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 84…85 °C (5mm)
Density: 1.71 g/cm³
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 84-85°C/5mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR