Pentafluorobenzamide ≥99%

Supplier: Matrix Scientific

Synonyms: 2,3,4,5,6-Pentafluorobenzamide

001543-25G 001543-5G 001543-100G
101781-934EA 35.38 USD
101781-934 101781-938 101781-936
Pentafluorobenzamide ≥99%
Pentafluorobenzamide
Formula: C₇H₂F₅NO
MW: 211.09 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 151 °C
MDL Number: MFCD00007971
CAS Number: 652-31-3
EINECS: 211-488-1

Order Now

Specification Test Results

Melting point 150°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR