2'-Fluoro-4'-hydroxyacetophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(2-Fluoro-4-hydroxyphenyl)ethanone
001479-25G
001479-5G
101781-778EA
53.85
USD
101781-778
101781-780
2'-Fluoro-4'-hydroxyacetophenone ≥97%
2'-Fluoro-4'-hydroxyacetophenone
Hazard code(s): Irritant
Formula:
C₈H₇FO₂ MW: 154.14 g/mol Melting Pt: 117…119 °C |
MDL Number:
MFCD00142713 CAS Number: 98619-07-9 |
Specification Test Results
Melting point | 117-119°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...