Decafluoro-5,6-bis(trifluoromethyl)cyclohexane ≥70%

Supplier: Matrix Scientific

Synonyms: Perfluoro-1,2-dimethylcyclohexane

001387-25G 001387-100G
101781-540EA 36.92 USD
101781-540 101781-542
Decafluoro-5,6-bis(trifluoromethyl)cyclohexane ≥70%
Decafluoro-5,6-bis(trifluoromethyl)cyclohexane

Hazard code(s): Irritant

Formula: C₈F₁₆
MW: 400.06 g/mol
Boiling Pt: 102 °C
Density: 1.867 g/cm³
MDL Number: MFCD00066612
CAS Number: 306-98-9
EINECS: 206-195-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Boiling point 102°C
Density 1.867
ND20 1.292

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR