4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥97%
Supplier: Matrix Scientific
001174-5G
001174-25G
101781-080EA
301.54
USD
101781-080
101781-078
4-Bromo-3,3,4,4-tetrafluoro-1-butene ≥97%
4-Bromo-3,3,4,4-tetrafluoro-1-butene
Hazard code(s): Irritant
Formula:
C₄H₃BrF₄ MW: 206.97 g/mol Boiling Pt: 55 °C Density: 1.627 g/cm³ |
MDL Number:
MFCD00042189 CAS Number: 18599-22-9 EINECS: 242-440-8 UN: 1993 ADR: 3,II |
Specification Test Results
Boiling point | 55°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...