Methyl-2-(trifluoromethoxy)benzoate ≥97%

Supplier: Matrix Scientific

001114-1G
101780-996EA 30.77 USD
101780-996
Methyl-2-(trifluoromethoxy)benzoate ≥97%
Methyl 2-(trifluoromethoxy)benzoate

Hazard code(s): Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Boiling Pt: 91…93 °C (15mm)
MDL Number: MFCD00052336
CAS Number: 148437-99-4

Order Now

Specification Test Results

Boiling point 91-93°C/15m

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR