Methyl-4-(trifluoromethoxy)benzoate ≥97%

Supplier: Matrix Scientific

001107-25G
101780-976EA 90.77 USD
101780-976
Methyl-4-(trifluoromethoxy)benzoate ≥97%
Methyl 4-(trifluoromethoxy)benzoate

Hazard code(s): Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
Boiling Pt: 88…90 °C (15mm Hg)
MDL Number: MFCD00041006
CAS Number: 780-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR