Perfluoro(methylcyclohexane) ≥90%
Supplier: Matrix Scientific
Synonyms:
FlutecR PP2
001095-100G
001095-25G
101780-940EA
63.08
USD
101780-940
101780-942
Perfluoro(methylcyclohexane) ≥90%
Perfluoro(methylcyclohexane)
Hazard code(s): Irritant
Formula:
C₇F₁₄ MW: 350.05 g/mol Boiling Pt: 76 °C Density: 1.787 g/cm³ |
MDL Number:
MFCD00001468 CAS Number: 355-02-2 EINECS: 206-573-5 |
Specification Test Results
Boiling point | 76°C |
Density | 1.787 |
ND20 | 1.3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...