ω-Bromo-4-fluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-(4-fluorophenyl)ethanoneα-Bromo-4-fluoroacetophenoneBromomethyl p-fluorophenyl ketone4-Fluorophenacyl bromide2-Bromo-4'-fluoroacetophenone2-Bromo-1-(4-fluorophenyl)ethan-1-one

001068-25G 001068-100G
101780-904EA 87.69 USD
101780-904 101780-902
ω-Bromo-4-fluoroacetophenone ≥97%
ω-Bromo-4-fluoroacetophenone

Hazard code(s): Corrosive, Lachrymator, Store in Freezer

Formula: C₈H₆BrFO
MW: 217.04 g/mol
Boiling Pt: 150 °C (12mm)
Melting Pt: 48…50 °C
MDL Number: MFCD00040830
CAS Number: 403-29-2
EINECS: 206-955-1

Order Now

Specification Test Results

Boiling point 150°C/12mm
Melting point 48-50°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR