Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Constant Climate Chamber with ICH-compliant Illumination, KBF P Series, BINDER

Supplier: BINDER INC

9020-0275 9020-0276 9020-0273
75796-928EA 31044.73 USD InStock
75796-928 75796-926 75796-924
Constant Climate Chamber with ICH-compliant Illumination, KBF P Series, BINDER
Testing Chambers Climatic Chambers
The expert for photostability testing

In terms of standar ds compliance, performance and functionality, the KBF P is a true KBF and is also equipped with ICH-compliant illumination. This provides clear and meaningful test results according to ICH guideline Q1B, Option 2. The variable position illumination cassettes with the unique BINDER Q1B Synergy Light ™ ensure homogeneous lighting conditions. Design: Inner glass door with smooth inner face and seals, access port with Ø 30 mm silicone plug on the left, includes stainless steel racks. Control: Electronic controlled APT.line™ preheating chamber, Screen controller with color LCD screen with 25 storable programs each with 100 sections for max. 500 program segments. Integrated electronic chart recorder, variety of options for the graphic display of process parameters and a real time clock.

Safety: Adjustable temperature safety device Class 3.1, DIN 12880 with visual and audible alarm.

Water Management: Water treatment with BINDER PURE AQUA SERVICE, Complete safety connection kit for water supply and drainage, up to 1m in height.

Humidity: Microprocessor controlled humidification and dehumidification system with a humidity range of 10 - 75% RH.

Connections: Ethernet interface for communication software (software optional). RS422 interface available as an option.

Certifications: UL Listed.

Ordering information: The KBF P 240 model is supplied with two stainless steel racks, the KBF P 720 model is supplied with three stainless stell racks.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

SERVICES


Services: Calibration and validation possible.

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty