Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Humidity Test Chambers, KBF Series, BINDER

Supplier: BINDER INC

9020-0264 9020-0271 9020-0272 9020-0168 9020-0150 9020-0184 9020-0139 9020-0108 9020-0184 9020-0176
75783-638EA 29010.95 USD InStock
75783-638 75796-922 75796-920 89182-848 89182-840 CA89182-870 89182-836 89182-832 89182-870 89182-858
Humidity Test Chambers, KBF Series, BINDER
Testing Chambers Climatic Chambers
The KBF series is ideal for reliable stability tests and precise maintenance of constant climatic conditions.

  • Inner glass door with smooth inner face and seals
  • Access port with Ø 30 mm silicone plug on the left
  • Includes 2 stainless steel racks
  • Adjustable temperature safety device Class 3.1, DIN 12880 with visual and audible alarm
  • Electronically controlled humidification and dehumidification system with a humidity range of 10 to 80% RH
  • Calibration and validation possible

Constant temperature and humidity values are the outstanding features of this range

Electronic controlled APT.line™ preheating chamber technology ensures long-term operational stability even under full load. Chambers feature state-of-the-art intuitive controller technology with time-segment and real-time programming with internal data recording. LCD color screen controller, with 25 storable programs each with 100 sections for a maximum of 500 program segments, includes a variety of options for the graphic display of process parameters and a real time clock.

Optional water treatment with Binder Pure Aqua Service, a complete safety connection kit for water supply and drainage, up to 1m in height. Ethernet interface for communication software (software optional). RS422 interface available as an option.

These constant climate chambers have a large reserve capacity and a variety of optional features. They also comply with all applicable guidelines with respect to programming and documentation requirements, such as ICH, FDA, GMP and GLP.

Certifications: UL Listed.

Delivery information: The KBF models are supplied with two stainless steel racks.

Order Now
 

ORDER

SPECIFICATIONS

ACCESSORIES

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty