Nessler reagent USP Test Solution (TS)
Supplier: Ricca Chemical
|
Danger
|
Synonyms:
Nota 1
, Potassium tetraiodomercurate
, Mercury (II) potassium iodide
, Potassium tetraiodomercurate – Potassium hydroxide solution
, Potassium mercuric Iodide
R5256000-500B
R5256000-500B
RC5256-16EA
141.01
USD
RC5256-16
RC525516
Nessler reagent USP Test Solution (TS)
Nessler's reagent
MW:
786.4 g/mol Boiling Pt: ∼100 °C (1013 hPa) Density: 1.2 g/cm³ (20 °C) |
MDL Number:
MFCD00049656 UN: 2922 ADR: 8,II |
Specification Test Results
Ammonia Sensitivity | Passes Test |
Appearance | Clear, light yellow liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...