Xylenol orange triturate 1% (w/w) in potassium nitrate

Supplier: Ricca Chemical
Warning

Synonyms: 2,2',2'',2'''-((((1,1-Dioxido-3H-benzo[c][1,2]oxathiole-3,3-diyl)bis(6-hydroxy-5-methyl-3,1-phenylene))bis(methylene))bis(azanetriyl))tetraacetic acid

R9395000-100H
RCR9395000100HEA 412.78 USD
RCR9395000100H
Xylenol orange triturate 1% (w/w) in potassium nitrate
Xylenol orange triturate 1% in potassium nitrate
Formula: C₃₁H₃₂N₂O₁₃S
MW: 672.67 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00014580
CAS Number: 1611-35-4
EINECS: 216-553-8
UN: 1486
ADR: 5.1,III

Order Now

Specification Test Results

Appearance Reddish-brown solid
Suitability for Use Purple (Excess Metal) - Yellow (Excess EDTA)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR