Ruthenium AA Standard, 1 ml = 1 mg Ru (1,000 ppm Ru)RuCl3 in 10% HCl, Ricca Chemical Company
Supplier: RICCA Chemical Company
RARU1KH0-100N
RCARU1KH-100EA
261.19
USD
RCARU1KH-100
Ruthenium AA Standard, 1 ml = 1 mg Ru (1,000 ppm Ru)RuCl3 in 10% HCl, Ricca Chemical Company
Standards
Cationic Standards
Ruthenium Standards