SP Bel-Art Magic Touch™ Round Floating Rack for 1.5 ml Microcentrifuge Tubes, Bel-Art Products, a part of SP
Supplier: Bel-Art
188480012
89198-988EA 49.93 USD
89198-988
SP Bel-Art Magic Touch™ Round Floating Rack for 1.5 ml Microcentrifuge Tubes, Bel-Art Products, a part of SP
Racks Tube Racks
Floating tube racks hold 44 microcentrifuge tubes and are convenient to use, and easy to store or transport.
  • Expanded polymer foam construction
  • Securely holds conical tubes
  • Floats in water, dry ice/alcohol slurries
Tubes insert easily into the expanded polymer foam which holds them secure yet allows them to come out without difficulty when needed.
These racks float in water, dry ice alcohol slurries or liquid nitrogen and fit neatly into Magic Touch 2.5L Handy and 4.0L N’Icer Ice Buckets.