Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Incubator Thermometer

Supplier: VWR International
VWR®
60810-2611 60170-4111 60170-4211 608102611
These are specific services for re-calibrating incubator thermometers for specified temperature points.

  • Celsius scale
  • Spirit-filled



 

ORDER

Expand All / Collapse All

Thermometers
Description Range Length Divisions Includes Immersion VWR Catalog Number Unit Price Quantity
Incubator Thermometer 24 to 57°C 175 mm (67/8") 0.5°C 35 mm (13/8") 89095-800 Case of 10 Retrieving
Each Retrieving

VWRCATALYST Re-Calibration Service (Service Only)
Description Range Length Divisions Includes Immersion VWR Catalog Number Unit Price Quantity
Customer Specified 1 Point Re-Calibration (0°C-100°C) ISO17025 NIST Certification, Customer specified 1 Temperature point Re-Calibration & no extra shipping fee 10186-026 Each Retrieving
Customer Specified 2-3 Points Re-Calibration (0°C-100°C) ISO17025 NIST Certification, Customer specified 2-3 Temperature points Re-Calibration & no extra shipping fee 10186-028 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: VWRCATALYST™ offers convenient and efficient re-calibration services for your liquid in glass thermometers with an industry leading 5-day average turn-around time from receipt of your instrument. Simply order the stated catalog no. based on how many calibration points are desired. You will receive a protective box to ship your instrument to VWR's certified calibration laboratory along with a work order form to specify your calibration points 0 to 100°C (32 to 212°F) and prepaid return shipping label. Upon receipt at the laboratory your instrument will be inspected for any defect under a microscope and the performance of your instrument will be compared to certified standards and corrections to one-tenth of the smallest division recorded on your signed calibration report. Your calibration report and instrument are carefully repacked then returned to you prepaid.

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty