    Sort by:
  • Search Within Results
Sudan black B
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"9"
Sort by:
Supplier:  VWR
Description:   A lipid stain used for demonstrating triglycerides in frozen sections. Also a histochemical stain for chromosomes, Golgi apparatus, and leucocyte gran...
SDS
Catalog Number: (TCS0113-025G)
Supplier:  TCI America
Description:   CAS Number: 4197-25-5 MDL Number: MFCD00006919 Molecular Formula: C29H24N6 Molecular Weight: 456.55 Form: Crystal Color: Deep Yellow Red Melting p...
SDS
Supplier:  MP Biomedicals
Description:   Soluble in ethanol; insoluble in water.
SDS
Supplier:  Spectrum Chemicals
Description:   SUDAN BLACK B 1KG
Small Business Enterprise
Supplier:  Alfa Aesar
Description:   Powder
SDS
Supplier:  Electron Microscopy Sciences
Description:   (Solvent Black 3) C29H24N6 M.W. 456.56 Solubility: Water-insoluble; Alcohol-soluble; 4.0% Cellosolve; 1.0% Glycol. A stain for fat in animal tissue....
Minority or Woman-Owned Business Enterprise
Supplier:  Acros Organics
Description:   Sudan Black B high purity biological stain, CAS Number: 4197-25-5, Molecular Formula: C29 H24 N6, Molecular weight: 456.54, Synonyms: C.I. 26150; Fat Black HB; Solvent Black 3, Form: Powder Solid, Color: Dark brown, Size: 25g
SDS
Catalog Number: (103463-882)
Supplier:  Acros Organics
Description:   Sudan Black B, certified, CAS number: 4197-25-5, Molecular Formula: C29H24N6, Molecular Weight: 456.55, Synonyms: C.I. 26150, Size: 10G
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 9
  1  2  Next