    Sort by:
  • Search Within Results
Acridine orange (hemizinc salt)
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"8"
Sort by:
Catalog Number: (97061-478)
Supplier:  VWR
Description:   A fluorescent stain for nucleic acids in agarose and polyacrylamide gels. Concentrations of 120 ?M will detect DNA at levels of 25-50 ng per band. Als...
SDS
Supplier:  MilliporeSigma
Description:   Cas Number: 10127-02-3, 25G
SDS
Supplier:  Alfa Aesar
Description:   Uses: Fluorescent stain for proteins. An RNA polymerase inhibitor
SDS
Catalog Number: (TCA0132-025G)
Supplier:  TCI America
Description:   CAS Number: 10127-02-3 MDL Number: MFCD00081043 Molecular Weight: 438.10 Form: Crystal
SDS
Supplier:  Biotium
Description:   stains dsdna green (525 nm) and rna or single stranded dna red (650 nm). the dye is cell permeable and its nucleic acid binding property has been used for cell-cycle studies. acridine orange has been used for the detction of microorganisms in cerebrospinal fluid.
Supplier:  Electron Microscopy Sciences
Description:   Electron Microscopy Sciences Acridine Orange Aqueous solution is a prepared, ready-to-use, high quality staining solutions for standard staining proce...
Minority or Woman-Owned Business Enterprise
Supplier:  Acros Organics
Description:   Size: 25GM. CAS Number: 10127-02-3.
SDS
Supplier:  Electron Microscopy Sciences
Description:   Electron Microscopy Sciences Acridine Orange Aqueous solution is a prepared, ready-to-use, high quality staining solutions for standard staining proce...
Minority or Woman-Owned Business Enterprise
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
???store.product.restricted???
This product has been blocked by your organization.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at 1-800-932-5000.
1 - 8  of 8