Would you like to log in again?
  • Criteria
  • Supplier
  • Suppliers found in search results
    Sort by:
  • Search Within Results
You Searched for: "cresol-red-sodium-salt-60-0-1-in-aqueous-solution"
SearchPresentationType-VERTICAL
SearchResultCount:"1"
Sort by:
Supplier:  Alfa Aesar
Description:   Transition interval (acid): pH 0.2 (red) to pH 1.8 (yellow); (alkaline) pH 7.2 (yellow) to pH 8.8 (red)