    Sort by:
  • Search Within Results
Isobutyl tiglate
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Click OK to update your results.
SearchResultCount:"1"
Sort by:
Catalog Number: (TCT1274-025ML)
Supplier:  TCI America
Description:   (stabilized with HQ) CAS Number: 61692-84-0 MDL Number: MFCD00016629 Molecular Formula: C9H16O2 Molecular Weight: 156.23 Purity/Analysis Method: <...