Would you like to log in again?
Corning® 96 Well Clear V-Bottom Polypropylene Not Treated Microplate
Supplier: Corning
3357
Corning 96-Well Polypropylene Microplates feature uniform skirt heights for greater robotic gripping surface
Their solvent resistant polypropylene provides compatibility with many common organic solvents (e.g., DMSO, ethanol, methanol). Sterile and certified DNase- and RNase-free. Total volume is 320µL; recommended working well volume is 75 to 200µL. Sold withouth lids; lids, sealing mats and tapes supplied separately.
ORDER
Expand All / Collapse All
Description Well Volume Color Sterility Well Shape Supplier No. VWR Catalog Number Unit Price Quantity
Corning® 96 Well Clear V-Bottom Polypropylene Not Treated Microplate 75 to 200 µL Clear Sterile V-Bottom 3357 29444-400 Case of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty